Assay Details
| Gene Symbol: Trnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242358.1 | NP_001229287.1 | 3 - 4 | 489 | 99 | |
| NM_001242360.1 | NP_001229289.1 | 3 - 4 | 424 | 99 | ||
| NM_027296.3 | NP_081572.1 | 3 - 4 | 424 | 99 | ||
| GenBank mRNA | AB063106.1 | - | 3 - 4 | 396 | 99 | |
| AK011764.1 | - | 3 - 4 | 440 | 99 | ||
| AK135791.1 | - | 3 - 4 | 444 | 99 | ||
| AK139764.1 | - | 3 - 4 | 405 | 99 | ||
| AK152739.1 | - | 3 - 4 | 460 | 99 | ||
| AK166804.1 | - | 3 - 4 | 434 | 99 | ||
| BC031764.1 | - | 3 - 4 | 390 | 4486420 | 99 | |
More Information
Set Membership: |
|

Trnt1
Genomic Map