Assay Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271397.1 | NP_001258326.1 | 7 - 8 | 2926 | 129 | |
| NR_073167.1 | 7 - 8 | 3107 | 129 | |||
| NR_073168.1 | 7 - 8 | 2922 | 129 | |||
| GenBank mRNA | AK029961.1 | - | 7 - 8 | 2639 | 129 | |
| AK147172.1 | - | 7 - 8 | 2662 | 129 | ||
| BC049166.1 | - | 7 - 8 | 2516 | 129 | ||
| BC058699.1 | - | 7 - 8 | 3082 | 129 | ||
| BC125575.1 | - | 7 - 8 | 2652 | 40130090 | 129 | |
| BC125579.1 | - | 7 - 8 | 2652 | 40130094 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11718 | BC058699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11718 | BC094941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11718 | BC125575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11718 | BC125579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11718 | AK147172.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684820 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87782 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106619 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167671 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11718 | BC058699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11718 | BC094941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11718 | BC125575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11718 | BC125579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11718 | AK147172.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07870490 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106619 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167671 | - | - |
| Whole Mouse Genome G4122A | A_52_P442722 | BC049166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442722 | BC058699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442722 | BC125575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442722 | BC125579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442722 | AK029961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366277 | BC049166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366277 | BC058699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366277 | BC094941.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366277 | BC125575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366277 | BC125579.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167671 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106619 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10509005 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10509002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10508991 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10508999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10967896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10491140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10509005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10452778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10691706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026598 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405079 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428951_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428950_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452974_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405074 | - | - |
| Mouse Genome 430 2.0 Array | 1428951_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428950_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452974_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452974_at | - | - |
| Mouse Genome 430 2.0 Array | 1428950_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428951_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70930_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70930_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300003540.mm.1 | NM_001271397.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003540.mm.1 | NR_073167.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003540.mm.1 | NR_073168.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003540.mm.1 | TC1300000571.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452974_at | - | - |
| Mouse Expression Set 430 | 1428950_PM_s_at | - | - |
| Mouse Expression Set 430 | 1452974_PM_at | - | - |
| Mouse Expression Set 430 | 1428950_s_at | - | - |
| Mouse Expression Set 430 | 1428951_at | - | - |
| Mouse Expression Set 430 | 1428951_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452974_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428950_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428950_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428951_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452974_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428951_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331667 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104590577 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106200671 | - | - |
| MouseWG-6 v2 Expression BeadChip | 100070204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104590577 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2627673 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243184 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222173 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104590577 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104590577 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243184 | - | - |

Nol8
Genomic Map