Assay Details
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029278.2 | NP_083554.2 | 5 - 6 | 851 | 121 | |
GenBank mRNA | AK011648.1 | - | 5 - 6 | 827 | 121 | |
AK078605.1 | - | 5 - 6 | 837 | 121 | ||
AK148605.1 | - | 5 - 6 | 810 | 121 | ||
AK161728.1 | - | 5 - 6 | 789 | 121 | ||
BC024998.1 | - | 5 - 6 | 772 | 5012070 | 121 | |
BC043043.1 | - | 5 - 6 | 794 | 5701430 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
