Assay Details
| Gene Symbol: Tti1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029282.1 | NP_083558.1 | 4 - 5 | 2826 | 79 | |
| GenBank mRNA | AF332067.1 | - | 4 - 5 | 2809 | 79 | |
| AF332068.1 | - | 4 - 5 | 2809 | 79 | ||
| AK128940.1 | - | 5 - 6 | 2876 | 79 | ||
| BC053067.1 | - | 4 - 5 | 2826 | 5698252 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tti1
Genomic Map