| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400027322.mm.1 |
TC0400003385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027322.mm.1 |
NM_001161823.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027322.mm.1 |
NM_001161823.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027322.mm.1 |
NM_172382.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027322.mm.1 |
NM_172382.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027316.mm.1 |
TC0400003385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027316.mm.1 |
NM_001161823.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027316.mm.1 |
NM_001161823.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027316.mm.1 |
NM_172382.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027316.mm.1 |
NM_172382.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027321.mm.1 |
TC0400003385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027321.mm.1 |
NM_001161823.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027321.mm.1 |
NM_001161823.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027321.mm.1 |
NM_172382.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027321.mm.1 |
NM_172382.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027320.mm.1 |
TC0400003385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027320.mm.1 |
NM_001161823.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027320.mm.1 |
NM_001161823.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027320.mm.1 |
NM_172382.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027320.mm.1 |
NM_172382.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027317.mm.1 |
TC0400003385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027317.mm.1 |
NM_001161823.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027317.mm.1 |
NM_001161823.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027317.mm.1 |
NM_172382.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027317.mm.1 |
NM_172382.2 |
Refseq |