Assay Details
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175413.3 | NP_780622.1 | 6 - 7 | 887 | 61 | |
GenBank mRNA | AK039277.1 | - | 6 - 7 | 884 | 61 | |
AK052159.1 | - | 6 - 7 | 839 | 61 | ||
AK079125.1 | - | 6 - 7 | 865 | 61 | ||
BC119529.2 | - | 6 - 7 | 836 | 40056120 | 61 | |
BC119530.1 | - | 6 - 7 | 837 | 40056121 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
