Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400024659.mm.1 |
TC0400003076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024659.mm.1 |
NM_177239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024659.mm.1 |
NM_177239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024665.mm.1 |
TC0400003076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024665.mm.1 |
NM_177239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024665.mm.1 |
NM_177239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024666.mm.1 |
TC0400003076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024666.mm.1 |
NM_177239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024666.mm.1 |
NM_177239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024670.mm.1 |
TC0400003076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024670.mm.1 |
NM_177239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024670.mm.1 |
NM_177239.2 |
Refseq |