Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400016537.mm.1 |
NM_178406.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016537.mm.1 |
NM_178406.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016537.mm.1 |
TC0400002018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016538.mm.1 |
NM_178406.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016538.mm.1 |
NM_178406.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016538.mm.1 |
TC0400002018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016539.mm.1 |
NM_178406.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016539.mm.1 |
NM_178406.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016539.mm.1 |
TC0400002018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016536.mm.1 |
NM_178406.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016536.mm.1 |
NM_178406.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016536.mm.1 |
TC0400002018.mm.1 |
MTA TC |