Assay Details
| Gene Symbol: 1700028K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175241.1 | NP_780450.1 | 5 - 6 | 540 | 106 | |
| NM_182745.2 | NP_877422.1 | 5 - 6 | 540 | 106 | ||
| GenBank mRNA | AK006463.1 | - | 5 - 6 | 540 | 106 | |
| CN835533.1 | - | 5 - 6 | 515 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158391 | NM_182745.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158391 | AK006463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158391 | AK029908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095424 | CN835533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095424 | AK032886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095424 | NM_175241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095424 | NM_182745.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095424 | AK006463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095425 | CN835533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095425 | AK032886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095425 | NM_175241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095425 | NM_182745.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095425 | AK006463.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62230 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166337 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152848 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134307 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137258 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095425 | CN835533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095425 | AK032886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095425 | NM_175241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095425 | NM_182745.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095425 | AK006463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158391 | NM_182745.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158391 | AK006463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158391 | AK029908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095424 | CN835533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095424 | AK032886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095424 | NM_175241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095424 | NM_182745.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095424 | AK006463.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23967471 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166337 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05687514 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05687525 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05687504 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05687493 | - | - |
| Whole Mouse Genome G4122A | A_52_P411076 | AK032886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P411076 | NM_175241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118244 | CN835533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118244 | AK032886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118244 | NM_175241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118244 | NM_182745.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118244 | AK006463.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136827 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137258 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05687514 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04224996 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04373004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04224997 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04224998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04889026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23967477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04441993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04454905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04984405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04984410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04361563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23967480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04139839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523873 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523865 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532103 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500009121.mm.1 | TC0500001079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009121.mm.1 | NM_175241.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009121.mm.1 | NM_175241.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009122.mm.1 | TC0500001079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009122.mm.1 | NM_175241.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009122.mm.1 | NM_175241.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009118.mm.1 | TC0500001079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009118.mm.1 | NM_175241.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009118.mm.1 | NM_175241.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009118.mm.1 | NM_182745.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009118.mm.1 | NM_182745.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009119.mm.1 | TC0500001079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009119.mm.1 | NM_182745.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009119.mm.1 | NM_182745.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 481345 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100510592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100510592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 840563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725531 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680815 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2680815 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225947 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725531 | - | - |
| mouse-6 v1.1 (Illumina) | scl076421.6_28-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0076421.1_95-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440068 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840563 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100510592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076421.1_95-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl076421.6_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100510592 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840563 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2680815 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2725531 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680815 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2725531 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2566312 | - | - |

1700028K03Rik
Genomic Map