Assay Details
| Gene Symbol: Gpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182805.2 | NP_877957.1 | 3 - 4 | 466 | 82 | |
| GenBank mRNA | AK008086.1 | - | 3 - 4 | 488 | 82 | |
| AK009457.1 | - | 3 - 4 | 488 | 82 | ||
| BC022625.1 | - | 3 - 4 | 467 | 4195300 | 82 | |
| BC026846.1 | - | 3 - 4 | 465 | 4194336 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpt
Genomic Map