Assay Details
| Gene Symbol: Gpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182805.2 | NP_877957.1 | 4 - 5 | 603 | 83 | |
| GenBank mRNA | AK008086.1 | - | 4 - 5 | 625 | 83 | |
| AK009457.1 | - | 4 - 5 | 625 | 83 | ||
| BC022625.1 | - | 4 - 5 | 604 | 4195300 | 83 | |
| BC026846.1 | - | 4 - 5 | 602 | 4194336 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpt
Genomic Map