Assay Details
| Gene Symbol: Gpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182805.2 | NP_877957.1 | 5 - 6 | 847 | 73 | |
| GenBank mRNA | BC022625.1 | - | 5 - 6 | 848 | 4195300 | 73 |
| BC026846.1 | - | 5 - 6 | 846 | 4194336 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350022 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458500 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134869 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134869 | - | - |
| Whole Mouse Genome G4122A | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | AK009457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | BC022625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | BC026846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | NM_182805.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426739 | AK008086.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102303 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118269 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362782 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362690 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362724 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362756 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350006 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424982 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426502_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453216_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424979 | - | - |
| Mouse Genome 430 2.0 Array | 1453216_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426502_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426502_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453216_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76282_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76282_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004411.mm.1 | NM_182805.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004411.mm.1 | TC1500000605.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426502_s_at | - | - |
| Mouse Expression Set 430 | 1426502_PM_s_at | - | - |
| Mouse Expression Set 430 | 1453216_at | - | - |
| Mouse Expression Set 430 | 1453216_PM_at | - | - |
| Murine Genome U74v2 Set | 132388_f_at | - | - |
| Murine Genome U74v2 Set | 137237_at | - | - |
| Murine Genome U74v2 Set | 132388_PM_f_at | - | - |
| Murine Genome U74v2 Set | 137237_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426502_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426502_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426502_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426502_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453216_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453216_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 556768 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2350270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683924 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2683924 | - | - |
| mouse-6 v1.1 (Illumina) | scl47787.2_58-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47787.2_58-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2832524 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2832524 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47787.2_58-S | - | - |

Gpt
Genomic Map