Assay Details
| Gene Symbol: 4931429L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183104.2 | NP_898927.2 | 5 - 6 | 684 | 97 | |
| GenBank mRNA | AK016488.1 | - | 5 - 6 | 682 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016488.1 transcripts.
Set Membership: |
|

4931429L15Rik
Genomic Map