Assay Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168386.2 | NP_001161858.1 | 2 - 3 | 668 | 73 | |
| NM_183115.5 | NP_898938.4 | 2 - 3 | 668 | 73 | ||
| GenBank mRNA | AK017973.1 | - | 2 - 3 | 431 | 73 | |
| BC085249.1 | - | 2 - 3 | 386 | 30726150 | 73 | |
| BU513278.1 | - | 3 - 4 | 776 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017973.1 transcripts.
Set Membership: |
|

Ccdc125
Genomic Map