Assay Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168386.2 | NP_001161858.1 | 6 - 7 | 985 | 102 | |
| NM_183115.5 | NP_898938.4 | 7 - 8 | 1063 | 102 | ||
| GenBank mRNA | AK017973.1 | - | 6 - 7 | 748 | 102 | |
| BC085249.1 | - | 7 - 8 | 781 | 30726150 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc125
Genomic Map