Assay Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243161.1 | NP_001230090.1 | 3 - 4 | 1212 | 69 | |
| NM_183142.4 | NP_898965.1 | 3 - 4 | 1086 | 69 | ||
| NR_040650.1 | 3 - 4 | 1216 | 69 | |||
| GenBank mRNA | AB288237.1 | - | 3 - 4 | 1206 | 69 | |
| AB288238.1 | - | 3 - 4 | 1210 | 69 | ||
| AK046475.1 | - | 3 - 4 | 1088 | 69 | ||
| AK134312.1 | - | 3 - 4 | 1187 | 69 | ||
| AK157746.1 | - | 3 - 4 | 1212 | 69 | ||
| BC061469.1 | - | 3 - 4 | 1195 | 6821552 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708993 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015217 | AK157746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015217 | BC061469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015217 | NM_183142.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015217 | AK046475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015217 | AK134312.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3418 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178148 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111429 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015217 | AK157746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015217 | BC061469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015217 | NM_183142.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015217 | AK046475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015217 | AK134312.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25709009 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178148 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01689644 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111429 | - | - |
| Whole Mouse Genome G4122A | A_52_P642541 | BC061469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361359 | AK157746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361359 | BC061469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361359 | NM_183142.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361359 | AK046475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361359 | AK134312.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126948 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178148 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06782036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06782022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25708992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25709024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07289703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07183480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25708999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25708996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25708993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07028993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25708995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06782035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25708994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25709016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06955880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06875068 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570765 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570766 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570768 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434187_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439976_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570764 | - | - |
| Mouse Genome 430 2.0 Array | 1434187_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434187_at | - | - |
| Mouse Genome 430 2.0 Array | 1439976_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439976_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 207958_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 207958_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001986.mm.1 | TC0800000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001982.mm.1 | TC0800000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001984.mm.1 | TC0800000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001985.mm.1 | TC0800000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001983.mm.1 | TC0800000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NM_001243161.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NM_001243161.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NM_183142.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NM_183142.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NR_040650.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | NR_040650.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001981.mm.1 | TC0800000268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434187_at | - | - |
| Mouse Expression Set 430 | 1434187_PM_at | - | - |
| Mouse Expression Set 430 | 1439976_PM_at | - | - |
| Mouse Expression Set 430 | 1439976_at | - | - |
| Murine Genome U74v2 Set | 169967_i_at | - | - |
| Murine Genome U74v2 Set | 169967_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439976_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434187_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439976_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434187_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 712021 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5570403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5570403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251311 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224712 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251311 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224712 | - | - |
| mouse-6 v1.1 (Illumina) | scl00207958.1_216-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0207958.5_71-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570403 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00207958.1_216-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0207958.5_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1224712 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2827701 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224712 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2827701 | - | - |

Alg11
Genomic Map