Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600013369.mm.1 |
TC0600001678.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600013369.mm.1 |
NM_183165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600013369.mm.1 |
NM_183165.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600013371.mm.1 |
TC0600001678.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600013371.mm.1 |
NM_183165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600013371.mm.1 |
NM_183165.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600013365.mm.1 |
TC0600001678.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600013365.mm.1 |
NM_183165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600013365.mm.1 |
NM_183165.3 |
Refseq |