Assay Details
| Gene Symbol: Mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183170.2 | NP_898993.1 | 4 - 5 | 577 | 69 | |
| GenBank mRNA | AK138901.1 | - | 4 - 5 | 603 | 69 | |
| AK151477.1 | - | 4 - 5 | 600 | 69 | ||
| AK152031.1 | - | 4 - 5 | 600 | 69 | ||
| AK168278.1 | - | 4 - 5 | 555 | 69 | ||
| AY013699.1 | - | 4 - 5 | 577 | 69 | ||
| BC051227.1 | - | 4 - 5 | 586 | 3470737 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953853 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481283 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25953842 | - | - |
| Whole Mouse Genome G4122A | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | AK152031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | AK168278.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | AY013699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | BC051227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483311 | NM_183170.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06950254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06950246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06950244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25953853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25953883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06950248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06924598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06924612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06950244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06950245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25953842 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579344 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434006_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579341 | - | - |
| Mouse Genome 430 2.0 Array | 1434006_at | - | - |
| Mouse Genome 430 2.0 Array | 1434006_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234384_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234384_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800018252.mm.1 | NM_183170.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800018252.mm.1 | NM_183170.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800018252.mm.1 | TC0800002402.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434006_at | - | - |
| Mouse Expression Set 430 | 1434006_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434006_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434006_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900497 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238114 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238114 | - | - |
| mouse-6 v1.1 (Illumina) | scl34694.5.9_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34694.5.9_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238114 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238114 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34694.5.9_23-S | - | - |

Mpv17l2
Genomic Map