Assay Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183281.2 | NP_899104.2 | 2 - 3 | 213 | 59 | |
| GenBank mRNA | AK009173.1 | - | 2 - 3 | 221 | 59 | |
| AK036849.1 | - | 2 - 3 | 240 | 59 | ||
| BC150914.1 | - | 1 - 2 | 366 | 59 | ||
| BC150924.1 | - | 1 - 2 | 366 | 59 | ||
| BC172084.1 | - | 1 - 2 | 366 | 9054370 | 59 | |
More Information
Set Membership: |
|

Tmem45a2
Genomic Map