Assay Details
| Gene Symbol: Tcerg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183289.3 | NP_899112.2 | 1 - 2 | 413 | 72 | |
| GenBank mRNA | AK017700.1 | - | 1 - 2 | 413 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017700.1 transcripts.
Set Membership: |
|

Tcerg1l
Genomic Map