Assay Details
| Gene Symbol: Timm17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011591.4 | NP_035721.1 | 3 - 4 | 238 | 148 | |
| GenBank mRNA | AF106621.1 | - | 3 - 4 | 192 | 148 | |
| AK003204.1 | - | 3 - 4 | 211 | 148 | ||
| AK009164.1 | - | 3 - 4 | 240 | 148 | ||
| AK011830.1 | - | 3 - 4 | 239 | 148 | ||
| AK049584.1 | - | 3 - 4 | 239 | 148 | ||
| AK150836.1 | - | 3 - 4 | 241 | 148 | ||
| AK153097.1 | - | 3 - 4 | 239 | 148 | ||
| BC008275.1 | - | 3 - 4 | 220 | 2811777 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32240781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32240782 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172299 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970920 | AK150836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970920 | AK153097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970920 | BC008275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970920 | NM_011591.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970920 | AK003204.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476782 | AK084534.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133198 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104114 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172299 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970920 | AK150836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970920 | AK153097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970920 | BC008275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970920 | NM_011591.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970920 | AK003204.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P238063 | NM_011591.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P238063 | AK049584.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338244 | AK150836.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338244 | AK153097.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338244 | BC008275.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338244 | NM_011591.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338244 | AK003204.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09227107 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104114 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09227116 | - | - |
| Whole Mouse Genome G4122A | A_51_P338244 | AK150836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338244 | AK153097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338244 | BC008275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338244 | NM_011591.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338244 | AK003204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P437321 | AK150836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P437321 | AK153097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P437321 | BC008275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P437321 | NM_011591.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P437321 | AK003204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474249 | AK150836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474249 | AK153097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474249 | BC008275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474249 | NM_011591.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474249 | AF106621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238063 | NM_011591.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238063 | AK049584.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100415 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104114 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09227116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14941587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14941582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14941588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14941583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32240782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32240781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14941587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14622047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14800421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14830877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14941589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15204846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598489 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460685_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438656_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598482 | - | - |
| Mouse Genome 430 2.0 Array | 1460685_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438656_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438656_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1460685_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21855_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21855_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00000597.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000597.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000599.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000599.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000600.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000600.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000604.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000604.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000607.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000607.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000605.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000605.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000602.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000602.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000601.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000601.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000608.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000608.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000606.mm.1 | NM_011591.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000606.mm.1 | TC0X00000076.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438656_PM_x_at | - | - |
| Mouse Expression Set 430 | 1460685_PM_at | - | - |
| Mouse Expression Set 430 | 1460685_at | - | - |
| Mouse Expression Set 430 | 1438656_x_at | - | - |
| Murine Genome U74v2 Set | 96668_PM_at | - | - |
| Murine Genome U74v2 Set | 96668_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438656_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460685_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460685_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438656_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438656_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460685_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438656_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460685_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 359400 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776304 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776304 | - | - |
| mouse-6 v1.1 (Illumina) | scl55067.6_230-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl55067.6_230-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2824012 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776304 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2824012 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl55067.6_230-S | - | - |

Timm17b
Genomic Map