Assay Details
| Gene Symbol: Nlrp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY364009.1 | - | 1848 | 114 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145739 | AY596202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145739 | DV652646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145739 | AK145355.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145739 | AY364009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145739 | NM_001048220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145740 | DV652646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145740 | AK145355.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145740 | NM_001048220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145740 | NM_001048219.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907436 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713852 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731425 | AK010864.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731425 | AK168268.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731425 | BC092288.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731425 | NM_001025586.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731425 | NM_001025587.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123554 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143578 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102086 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145740 | DV652646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145740 | AK145355.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145740 | NM_001048220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145740 | NM_001048219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145739 | AY596202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145739 | DV652646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145739 | AK145355.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145739 | AY364009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145739 | NM_001048220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P100391 | AY596202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P100391 | DV652646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P100391 | AK145355.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P100391 | AY364009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P100391 | NM_001048220.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06230913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25014269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06536121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25014252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06640393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06190676 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551263 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551262 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456185_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551252 | - | - |
| Mouse Genome 430 2.0 Array | 1456185_at | - | - |
| Mouse Genome 430 2.0 Array | 1456185_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233001_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233001_PM_at | - | - |
| Mouse Expression Set 430 | 1456185_at | - | - |
| Mouse Expression Set 430 | 1456185_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456185_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456185_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_3099303 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3099303 | - | - |

Nlrp9a
Genomic Map