Assay Details
Gene Symbol: Vmn1r69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145842.3 | NP_665841.1 | 1 | 1562 | 84 | |
GenBank mRNA | AF394954.1 | - | 1 | 1561 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124263 | AF394954.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124263 | NM_145842.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124263 | AF394954.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124263 | NM_145842.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279938 | AF394954.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279938 | NM_145842.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966579 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24966613 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06122280 | - | - |
Whole Mouse Genome G4122A | A_51_P279938 | AF394954.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279938 | NM_145842.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130127 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06122280 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183498 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183488 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06201044 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183485 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183481 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183483 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06183500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24966577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06121703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06593477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06593474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06113250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06083520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06593470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06592153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06592155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06201042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06183488 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24966619 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10559973 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559971 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559972 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10559970 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700020750.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020750.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020750.mm.1 | NM_145842.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020746.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020746.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020746.mm.1 | NM_145842.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020745.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020745.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020745.mm.1 | NM_145842.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020751.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020751.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020751.mm.1 | NM_145842.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020747.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020747.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020747.mm.1 | NM_145842.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020749.mm.1 | TC0700002314.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020749.mm.1 | NM_145842.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020749.mm.1 | NM_145842.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 390887 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5890008 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005890008 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233229 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1233229 | - | - |
mouse-6 v1.1 (Illumina) | scl0252904.1_239-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5890008 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0252904.1_239-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890008 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1233229 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233229 | - | - |
