Assay Details
| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011348.2 | NP_035478.2 | 17 | 115 | ||
| GenBank mRNA | AK032603.1 | - | 1 | 115 | ||
| BE947109.1 | - | 1 | 205 | 115 | ||
More Information
Set Membership: |
|

Sema3e
Genomic Map