Assay Details
Gene Symbol: Gm5115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK086292.1 | - | 2751 | 126 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170284 | AK086292.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158816 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158816 | - | - |
Whole Mouse Genome G4122A | A_51_P219139 | AK086292.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135797 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25364716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06421026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05913464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05978424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06169067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05978431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06421025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06460184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06106794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06462082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25364770 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10565859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10565860 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10565861 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10565858 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6420072 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6420072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240400 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240400 | - | - |
mouse-6 v1.1 (Illumina) | scl30992.1.1192_73-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420072 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30992.1.1192_73-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2767162 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2767162 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30992.1.1192_73-S | - | - |
