Assay Details
| Gene Symbol: Eif5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177586.5 | NP_808254.1 | 2 - 3 | 299 | 86 | |
| GenBank mRNA | AK036127.1 | - | 2 - 3 | 301 | 86 | |
| AK052412.1 | - | 2 - 3 | 243 | 86 | ||
| AK082214.1 | - | 2 - 3 | 216 | 86 | ||
| AY205262.1 | - | 2 - 3 | 299 | 86 | ||
| AY205263.1 | - | 2 - 3 | 299 | 86 | ||
| AY205264.1 | - | 2 - 3 | 299 | 86 | ||
| AY205265.1 | - | 2 - 3 | 299 | 86 | ||
| BC106843.2 | - | 2 - 3 | 229 | 40045849 | 86 | |
| BC106844.1 | - | 2 - 3 | 229 | 40045850 | 86 | |
More Information
Set Membership: |
|

Eif5a2
Genomic Map