Assay Details
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 9 - 10 | 1232 | 97 | |
AK040344.1 | - | 7 - 8 | 1010 | 97 | ||
AK052103.1 | - | 7 - 8 | 960 | 97 | ||
AK085336.1 | - | 8 - 9 | 1106 | 97 | ||
AK085362.1 | - | 9 - 10 | 1271 | 97 | ||
AK086462.1 | - | 9 - 10 | 1264 | 97 | ||
BC054120.1 | - | 9 - 10 | 1217 | 6506658 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
