Assay Details
| Gene Symbol: Dhrs13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183286.2 | NP_899109.2 | 4 - 5 | 740 | 60 | |
| GenBank mRNA | AK011939.1 | - | 4 - 5 | 715 | 60 | |
| AK046465.1 | - | 3 - 4 | 787 | 60 | ||
| BC115881.1 | - | 3 - 4 | 585 | 40088339 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
metabolic process
oxidation-reduction process
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity
metabolic process
oxidation-reduction process
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941037 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013928 | AK046465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013928 | BC115881.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013928 | NM_183286.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013928 | AK011939.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231484 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151385 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111764 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013928 | AK046465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013928 | BC115881.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013928 | NM_183286.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013928 | AK011939.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27941055 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111764 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151385 | - | - |
| Whole Mouse Genome G4122A | A_51_P487941 | AK011939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487941 | AK046465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487941 | BC115881.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487941 | NM_183286.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122278 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111764 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09562300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09427100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09562301 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09013525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09000887 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09427098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09000886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09000888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09562302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09000898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27941030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09356348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09000890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09219175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09427100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09000891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09013525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09000887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09062647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09562302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941042 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379012 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452902_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379006 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388732 | - | - |
| Mouse Genome 430 2.0 Array | 1452902_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452902_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70451_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70451_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100010465.mm.1 | NM_183286.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010465.mm.1 | NM_183286.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010465.mm.1 | TC1100001139.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010466.mm.1 | NM_183286.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010466.mm.1 | NM_183286.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010466.mm.1 | TC1100001139.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452902_PM_at | - | - |
| Mouse Expression Set 430 | 1452902_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452902_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452902_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 620528 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 780132 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 780132 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780132 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770270 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770270 | - | - |
| mouse-6 v1.1 (Illumina) | scl41221.5_572-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780132 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41221.5_572-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780132 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2770270 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770270 | - | - |

Dhrs13
Genomic Map