Assay Details
| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182993.2 | NP_892038.2 | 5 - 6 | 799 | 50 | |
| GenBank mRNA | AK159064.1 | - | 5 - 6 | 772 | 50 | |
| BC028938.1 | - | 3 - 4 | 238 | 50 | ||
| BC054462.1 | - | 3 - 4 | 581 | 6494471 | 50 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc17a7
Genomic Map