Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300001219.mm.1 |
NM_183014.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001219.mm.1 |
NM_183014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001219.mm.1 |
TC1300000186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300001220.mm.1 |
NM_183014.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001220.mm.1 |
NM_183014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001220.mm.1 |
TC1300000186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300001223.mm.1 |
NM_183014.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001223.mm.1 |
NM_183014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300001223.mm.1 |
TC1300000186.mm.1 |
MTA TC |