Assay Details
Gene Symbol: Zfp184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183014.1 | NP_898835.1 | 5 - 6 | 648 | 132 | |
GenBank mRNA | AK036164.1 | - | 5 - 6 | 664 | 132 | |
BC053084.1 | - | 5 - 6 | 648 | 5708591 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875420 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875422 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875421 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074246 | BC053084.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074246 | NM_183014.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074246 | AK036164.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30508 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150466 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325273 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380033 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787204 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173810 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212235 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380734 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465517 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783834 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367821 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55680 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132369 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139091 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135505 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074246 | BC053084.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074246 | NM_183014.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074246 | AK036164.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875415 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875398 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28875400 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02644753 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07807106 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139091 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135505 | - | - |
Whole Mouse Genome G4122A | A_51_P143528 | BC053084.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P143528 | NM_183014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P143528 | AK036164.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108988 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113283 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135505 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139091 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11082045 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11082042 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11060727 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11060740 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11082046 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11060741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875422 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875421 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28875420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28875449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10703214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11060743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875397 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10928500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10471868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11082046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11025418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10848056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11082039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10703224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10525419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10791482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11060740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10525420 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11060731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11060739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875394 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10729511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11053940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10594725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10469931 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28875402 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10403970 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403966 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403965 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403972 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403968 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403971 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403967 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441997_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403964 | - | - |
Mouse Genome 430 2.0 Array | 1441997_at | - | - |
Mouse Genome 430 2.0 Array | 1441997_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193452_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193452_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001231.mm.1 | NM_183014.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001231.mm.1 | NM_183014.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001231.mm.1 | TC1300000186.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001232.mm.1 | NM_183014.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001232.mm.1 | NM_183014.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001232.mm.1 | TC1300000186.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001230.mm.1 | NM_183014.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001230.mm.1 | NM_183014.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001230.mm.1 | TC1300000186.mm.1 | MTA TC |
Mouse Expression Set 430 | 1441997_PM_at | - | - |
Mouse Expression Set 430 | 1441997_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441997_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441997_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 871711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102060541 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3830736 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102060541 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101770338 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003830736 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257607 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257607 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2433174 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2416545 | - | - |
mouse-6 v1.1 (Illumina) | scl45004.6_40-S | - | - |
mouse-6 v1.1 (Illumina) | scl00193452.2_269-S | - | - |
mouse-6 v1.1 (Illumina) | scl44227.2.1_53-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102060541 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101770338 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3830736 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00193452.2_269-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830736 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101770338 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102060541 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257607 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2416545 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257607 | - | - |
