Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700008712.mm.1 |
NM_183086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008712.mm.1 |
NM_183086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008712.mm.1 |
TC1700000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_001146211.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_001146211.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_001146212.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_001146212.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_183086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
NM_183086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008727.mm.1 |
TC1700000912.mm.1 |
MTA TC |