Assay Details
| Gene Symbol: Rims4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183023.1 | NP_898844.1 | 4 - 5 | 448 | 57 | |
| GenBank mRNA | AK134613.1 | - | 4 - 5 | 694 | 57 | |
| AY326955.1 | - | 4 - 5 | 448 | 57 | ||
| BC115475.1 | - | 4 - 5 | 447 | 57 | ||
| BC115476.1 | - | 2 - 3 | 57 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rims4
Genomic Map