Assay Details
| Gene Symbol: Trmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164559.1 | NP_001158031.1 | 9 - 10 | 1223 | 60 | |
| NM_001164560.1 | NP_001158032.1 | 9 - 10 | 1147 | 60 | ||
| NM_198020.2 | NP_932137.2 | 9 - 10 | 1250 | 60 | ||
| GenBank mRNA | AK032945.1 | - | 7 - 8 | 2683 | 60 | |
| AK084775.1 | - | 9 - 10 | 1189 | 60 | ||
| AK155251.1 | - | 9 - 10 | 1216 | 60 | ||
| AK159467.1 | - | 9 - 10 | 1188 | 60 | ||
| AK170412.1 | - | 9 - 10 | 1112 | 60 | ||
| AK171002.1 | - | 9 - 10 | 1215 | 60 | ||
| AK178569.1 | - | 2 - 3 | 126 | 60 | ||
| BC027404.1 | - | 9 - 10 | 1219 | 4950980 | 60 | |
More Information
Set Membership: |
|

Trmt1
Genomic Map