Assay Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176837.2 | NP_789807.1 | 9 - 10 | 1356 | 75 | |
| GenBank mRNA | AK014728.1 | - | 9 - 10 | 1304 | 75 | |
| AK078522.1 | - | 4 - 5 | 408 | 75 | ||
| AK087800.1 | - | 9 - 10 | 1334 | 75 | ||
| AK162202.1 | - | 5 - 6 | 599 | 75 | ||
| BC030858.1 | - | 9 - 10 | 1356 | 4194996 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap18
Genomic Map