Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 19 - 20 | 2239 | 68 | |
| NM_001285993.1 | NP_001272922.1 | 18 - 19 | 2140 | 68 | ||
| NM_028162.4 | NP_082438.3 | 18 - 19 | 2173 | 68 | ||
| GenBank mRNA | AK154975.1 | - | 19 - 20 | 2199 | 68 | |
| AK163366.1 | - | 18 - 19 | 2203 | 68 | ||
| AK220251.1 | - | 18 - 19 | 2140 | 68 | ||
| BC043113.1 | - | 18 - 19 | 1989 | 6407234 | 68 | |
| BC098328.1 | - | 13 - 14 | 1519 | 30357143 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d5
Genomic Map