Assay Details
| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007562.2 | NP_031588.2 | 2 - 3 | 200 | 92 | |
| GenBank mRNA | AK087567.1 | - | 2 - 3 | 194 | 92 | |
| BY146829.1 | - | 2 - 3 | 200 | 92 | ||
| U88064.1 | - | 2 - 3 | 147 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bnc1
Genomic Map