Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026267.mm.1 |
TC0700002964.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026267.mm.1 |
NM_197991.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026267.mm.1 |
NM_197991.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026272.mm.1 |
TC0700002964.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026272.mm.1 |
NM_197991.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026272.mm.1 |
NM_197991.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026271.mm.1 |
TC0700002964.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026271.mm.1 |
NM_197991.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026271.mm.1 |
NM_197991.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026266.mm.1 |
TC0700002964.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026266.mm.1 |
NM_197991.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026266.mm.1 |
NM_197991.2 |
Refseq |