Assay Details
Gene Symbol: Med16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163276.1 | NP_001156748.1 | 3 - 4 | 390 | 72 | |
NM_198107.2 | NP_932775.2 | 3 - 4 | 393 | 72 | ||
GenBank mRNA | AK042340.1 | - | 3 - 4 | 355 | 72 | |
AK138125.1 | - | 3 - 4 | 377 | 72 | ||
AK149345.1 | - | 3 - 4 | 374 | 72 | ||
BC057056.1 | - | 3 - 4 | 347 | 6813010 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
