Assay Details
Gene Symbol: Prp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031499.2 | NP_113687.2 | 62 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685719 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24853952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24853948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24853947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24853949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24853968 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06072722 | - | - |
Whole Mouse Genome G4122A | A_51_P219396 | M19419.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P383857 | NM_031499.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06072722 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05543008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05197716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05180820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05543008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24853968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05180822 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10542234 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542240 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542232 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542236 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542233 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450837_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10542231 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10542239 | - | - |
Mouse Genome 430 2.0 Array | 1450837_at | - | - |
Mouse Genome 430 2.0 Array | 1450837_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600012602.mm.1 | TC0600001559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012602.mm.1 | NM_031499.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012602.mm.1 | NM_031499.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012603.mm.1 | TC0600001559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012603.mm.1 | NM_031499.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012603.mm.1 | NM_031499.2 | Refseq |
Mouse Expression Set 430 | 1450837_PM_at | - | - |
Mouse Expression Set 430 | 1450837_at | - | - |
Murine Genome U74v2 Set | 93650_PM_i_at | - | - |
Murine Genome U74v2 Set | 93651_r_at | - | - |
Murine Genome U74v2 Set | 93650_i_at | - | - |
Murine Genome U74v2 Set | 93651_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1450837_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450837_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450837_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450837_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 901462 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2690500 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2690500 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002690500 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218867 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1218867 | - | - |
mouse-6 v1.1 (Illumina) | scl0083380.1_28-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2690500 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0083380.1_28-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690500 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2936574 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218867 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2936574 | - | - |
