Assay Details
| Gene Symbol: Hist1h3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006742.1 | - | 1 | 408 | 136 | |
| AK018952.1 | - | 1 | 429 | 136 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883375 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883378 | - | - |
| Whole Mouse Genome G4122A | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518621 | BC115816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518621 | AK006742.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133954 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682953 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682943 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10682944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28883376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28883378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28883380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408248 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408246 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403941 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404028 | - | - |
| Mouse Genome 430 2.0 Array | 1422948_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422947_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422948_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422947_at | - | - |
| Mouse Expression Set 430 | 1427864_PM_at | - | - |
| Mouse Expression Set 430 | 1460314_s_at | - | - |
| Mouse Expression Set 430 | 1427864_at | - | - |
| Mouse Expression Set 430 | 1460314_PM_s_at | - | - |
| Murine Genome U74v2 Set | 96416_f_at | - | - |
| Murine Genome U74v2 Set | 93023_f_at | - | - |
| Murine Genome U74v2 Set | 93023_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94756_at | - | - |
| Murine Genome U74v2 Set | 94756_PM_at | - | - |
| Murine Genome U74v2 Set | 96416_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2945694 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945694 | - | - |

Hist1h3a
Genomic Map