Assay Details
| Gene Symbol: Atp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007507.2 | NP_031533.2 | 3 - 4 | 205 | 139 | |
| GenBank mRNA | BC028438.1 | - | 3 - 4 | 205 | 1332765 | 139 |
| BC089550.1 | - | 3 - 4 | 197 | 30380642 | 139 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971890 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2168663 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2168664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16154 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150852 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2168663 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149719 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2168664 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301289 | NM_007507.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301289 | BC028438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301289 | BC089550.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301289 | S52977.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23971885 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150852 | - | - |
| Whole Mouse Genome G4122A | A_51_P301289 | NM_007507.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301289 | BC028438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301289 | BC089550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301289 | S52977.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136143 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136692 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107792 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372700 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372709 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372687 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372702 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04874061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23971893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04982630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372702 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532168 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434053_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450640_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422525_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532164 | - | - |
| Mouse Genome 430 2.0 Array | 1450640_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1434053_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450640_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422525_at | - | - |
| Mouse Genome 430 2.0 Array | 1434053_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422525_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11958_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11958_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500026027.mm.1 | TC0500002944.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026027.mm.1 | NM_007507.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026027.mm.1 | NM_007507.2 | Refseq |
| Mouse Expression Set 430 | 1422525_PM_at | - | - |
| Mouse Expression Set 430 | 1450640_x_at | - | - |
| Mouse Expression Set 430 | 1450640_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422525_at | - | - |
| Mouse Expression Set 430 | 1434053_PM_x_at | - | - |
| Mouse Expression Set 430 | 1434053_x_at | - | - |
| Murine Genome U74v2 Set | 102561_PM_at | - | - |
| Murine Genome U74v2 Set | 102561_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422525_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450640_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434053_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434053_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450640_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422525_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450640_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422525_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450640_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434053_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434053_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422525_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6220047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776900 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776900 | - | - |
| mouse-6 v1.1 (Illumina) | scl011958.2_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011958.2_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776899 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776899 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl011958.2_29-S | - | - |

Atp5k
Genomic Map