Assay Details
| Gene Symbol: Cflar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289704.2 | NP_001276633.1 | 4 - 5 | 766 | 102 | |
| NM_001293804.1 | NP_001280733.1 | 4 - 5 | 749 | 102 | ||
| NM_001293805.1 | NP_001280734.1 | 2 - 3 | 242 | 102 | ||
| NM_009805.4 | NP_033935.2 | 4 - 5 | 1038 | 102 | ||
| NM_207653.5 | NP_997536.1 | 4 - 5 | 1038 | 102 | ||
| GenBank mRNA | AK143812.1 | - | 4 - 5 | 1031 | 102 | |
| AK152515.1 | - | 4 - 5 | 733 | 102 | ||
| AK161120.1 | - | 2 - 3 | 245 | 102 | ||
| BC029223.1 | - | 4 - 5 | 1007 | 4460554 | 102 | |
| BC085105.1 | - | 4 - 5 | 761 | 102 | ||
| U97076.1 | - | 4 - 5 | 704 | 102 | ||
| Y14041.1 | - | 3 - 4 | 616 | 102 | ||
| Y14042.1 | - | 3 - 4 | 616 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293804.1, NM_001293805.1 transcripts.
Set Membership: |
|

Cflar
Genomic Map