Assay Details
| Gene Symbol: Krt6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010669.2 | NP_034799.2 | 1 - 2 | 576 | 199 | |
| GenBank mRNA | AK137688.1 | - | 1 - 2 | 575 | 199 | |
| BC119554.1 | - | 534 | 40057432 | 199 | ||
| BC119555.1 | - | 534 | 40057433 | 199 | ||
| BC122884.1 | - | 1 - 2 | 533 | 199 | ||
| BC124556.1 | - | 1 - 2 | 533 | 199 | ||
| BC139807.1 | - | 534 | 40057436 | 199 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101935 | DV553143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101935 | NM_010669.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101935 | AK009241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101935 | AK137688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101935 | DV553143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101935 | NM_010669.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101935 | AK009241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101935 | AK137688.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493286 | - | - |
| Whole Mouse Genome G4122A | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC139807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC119554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC119555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC122884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126275 | BC124556.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101935 | DV553143.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101935 | NM_010669.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101935 | AK009241.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101935 | AK137688.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12312730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12312748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12312751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12312715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12000052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432779 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422588_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432774 | - | - |
| Mouse Genome 430 2.0 Array | 1422588_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422588_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16688_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16688_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018657.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018657.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018657.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018662.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018662.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018662.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018661.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018661.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018661.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018655.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018655.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018655.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018658.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018658.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018658.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018659.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018659.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018659.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018663.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018663.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018663.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018665.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018665.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018665.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018656.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018656.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018656.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018664.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018664.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018664.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018660.mm.1 | NM_010669.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018660.mm.1 | NM_010669.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018660.mm.1 | TC1500002268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422588_PM_at | - | - |
| Mouse Expression Set 430 | 1422588_at | - | - |
| Murine Genome U74v2 Set | 94316_PM_at | - | - |
| Murine Genome U74v2 Set | 94316_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422588_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422588_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422588_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422588_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 695928 | - | - |
| ABI Mouse Genome Survey Microarray | 744954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940088 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4070725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235713 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235713 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594736 | - | - |
| mouse-6 v1.1 (Illumina) | scl0016688.1_300-S | - | - |
| mouse-6 v1.1 (Illumina) | scl016688.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070725 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016688.1_300-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016688.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940088 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235713 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594736 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235713 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594736 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0016688.1_300-S | - | - |

Krt6b
Genomic Map