Assay Details
| Gene Symbol: Atp5j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020582.2 | NP_065607.1 | 2 - 3 | 197 | 134 | |
| GenBank mRNA | AB030192.1 | - | 2 - 3 | 198 | 134 | |
| AK002519.1 | - | 2 - 3 | 147 | 134 | ||
| AK003235.1 | - | 2 - 3 | 135 | 134 | ||
| AK003817.1 | - | 2 - 3 | 147 | 134 | ||
| AK013130.1 | - | 2 - 3 | 147 | 134 | ||
| AK151737.1 | - | 2 - 3 | 146 | 134 | ||
| AK168095.1 | - | 2 - 3 | 147 | 134 | ||
| BC029226.1 | - | 1 - 2 | 137 | 2651401 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081510 | AK013130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081510 | AK151737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081510 | AK168095.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081510 | NM_020582.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081510 | AB030192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312356 | AK085304.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125986 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132219 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081510 | AK013130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081510 | AK151737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081510 | AK168095.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081510 | NM_020582.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081510 | AB030192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118906 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224216 | AK013130.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224216 | AK151737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224216 | AK168095.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224216 | BC029226.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224216 | NM_020582.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164797 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01243988 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132219 | - | - |
| Whole Mouse Genome G4122A | A_51_P224216 | AK013130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224216 | AK151737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224216 | AK168095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224216 | BC029226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224216 | NM_020582.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120387 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435149 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435141 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435148 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04435143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04507532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04745576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04152427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04627853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04741747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04248294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04507533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04500366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04508898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04988276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04435148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04508897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04435139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04500367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04621033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04708107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04435149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04435144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04886181 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535648 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535649 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535651 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443495_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435395_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416269_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535647 | - | - |
| Mouse Genome 430 2.0 Array | 1443495_at | - | - |
| Mouse Genome 430 2.0 Array | 1416269_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435395_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1435395_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1443495_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416269_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57423_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57423_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032948.mm.1 | TC0500003591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032948.mm.1 | NM_020582.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032948.mm.1 | NM_020582.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032951.mm.1 | TC0500003591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032951.mm.1 | NM_020582.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032951.mm.1 | NM_020582.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032959.mm.1 | TC0500003591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032959.mm.1 | NM_020582.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032959.mm.1 | NM_020582.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032950.mm.1 | TC0500003591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032950.mm.1 | NM_020582.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032950.mm.1 | NM_020582.2 | Refseq |
| Mouse Expression Set 430 | 1435395_PM_s_at | - | - |
| Mouse Expression Set 430 | 1416269_PM_at | - | - |
| Mouse Expression Set 430 | 1435395_s_at | - | - |
| Mouse Expression Set 430 | 1416269_at | - | - |
| Mouse Expression Set 430 | 1443495_at | - | - |
| Mouse Expression Set 430 | 1443495_PM_at | - | - |
| Murine Genome U74v2 Set | 93114_at | - | - |
| Murine Genome U74v2 Set | 93114_PM_at | - | - |
| Murine Genome U74v2 Set | 168278_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168278_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416269_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416269_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435395_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435395_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435395_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435395_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416269_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416269_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443495_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443495_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213914 | - | - |
| mouse-6 v1.1 (Illumina) | scl057423.2_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl057423.2_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213914 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213914 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl057423.2_11-S | - | - |

Atp5j2
Genomic Map