Assay Details
Gene Symbol: Sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024171.2 | NP_077133.1 | 3 - 4 | 298 | 94 | |
GenBank mRNA | AK004505.1 | - | 3 - 4 | 299 | 94 | |
AK008468.2 | - | 741 | 94 | |||
AK008739.1 | - | 3 - 4 | 302 | 94 | ||
BC002089.1 | - | 3 - 4 | 270 | 3487410 | 94 | |
BC081445.1 | - | 3 - 4 | 266 | 6412192 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051962 | BC002089.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051962 | BC081445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051962 | NM_024171.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051962 | AK004505.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P112593 | AK008739.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P112593 | NM_024171.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P112593 | AK004505.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334714 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310625 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332937 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728107 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769100 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65712 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64840 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167454 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P112593 | AK008739.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P112593 | NM_024171.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P112593 | AK004505.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051962 | BC002089.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051962 | BC081445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051962 | NM_024171.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051962 | AK004505.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045481 | BC002089.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894918 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894901 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894912 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22894919 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167454 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05227363 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05227373 | - | - |
Whole Mouse Genome G4122A | A_52_P93886 | BC002089.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93886 | BC081445.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93886 | AK008739.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93886 | NM_024171.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93886 | AK004505.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106229 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112593 | AK008739.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112593 | NM_024171.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112593 | AK004505.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126964 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167454 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03605512 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03605509 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03605517 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03605516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22895008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22894984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04053289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22895009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03541556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22895000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22895015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04053295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04053293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03628701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22895006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894919 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894905 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03766453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03440844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03364158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04053294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22895007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03627882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22894901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03605512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03440848 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10426890 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10411455 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10504833 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417083_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10411454 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10426889 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10504831 | - | - |
Mouse Genome 430 2.0 Array | 1417083_at | - | - |
Mouse Genome 430 2.0 Array | 1417083_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66212_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66212_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400003852.mm.1 | NM_024171.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003852.mm.1 | NM_024171.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003852.mm.1 | TC0400000523.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400003854.mm.1 | NM_024171.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003854.mm.1 | NM_024171.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003854.mm.1 | TC0400000523.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400003856.mm.1 | NM_024171.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003856.mm.1 | NM_024171.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003856.mm.1 | TC0400000523.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400003855.mm.1 | NM_024171.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003855.mm.1 | NM_024171.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400003855.mm.1 | TC0400000523.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417083_at | - | - |
Mouse Expression Set 430 | 1417083_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417083_at | - | - |
Mouse Genome 430A 2.0 Array | 1417083_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417083_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417083_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 895902 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 840113 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 840113 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540687 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540687 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000840113 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2609807 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770628 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2770628 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2609807 | - | - |
mouse-6 v1.1 (Illumina) | scl066212.2_13-S | - | - |
mouse-6 v1.1 (Illumina) | scl0066212.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540687 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 840113 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066212.2_13-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066212.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840113 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540687 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2609809 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2609807 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2609809 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2609807 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066212.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066212.2_13-S | - | - |
