Assay Details
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025440.2 | NP_079716.1 | 2 - 3 | 337 | 162 | |
GenBank mRNA | AB049949.1 | - | 2 - 3 | 395 | 162 | |
AK005206.1 | - | 2 - 3 | 338 | 162 | ||
AK019243.1 | - | 2 - 3 | 283 | 162 | ||
BC024656.1 | - | 2 - 3 | 303 | 3986314 | 162 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446630 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446629 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446632 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446633 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446634 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166538 | AK005206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166538 | NM_025440.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166538 | AK019243.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166538 | BC024656.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312410 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102174 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166538 | AK005206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166538 | NM_025440.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166538 | AK019243.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166538 | BC024656.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164425 | AK005206.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164425 | NM_025440.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164425 | AK019243.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164425 | BC024656.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446639 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446658 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29446668 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102174 | - | - |
Whole Mouse Genome G4122A | A_51_P164425 | AK005206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164425 | NM_025440.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164425 | AK019243.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164425 | BC024656.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111221 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102174 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11579059 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11579044 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11579055 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11579048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446630 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446629 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446634 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446632 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29446633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29446591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11280966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11359811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11734618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11578230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11280968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11579055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11280964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11330878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11382405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11696681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11573452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11807966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11280965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29446646 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10417840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417839 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448869_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10417836 | - | - |
Mouse Genome 430 2.0 Array | 1448869_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1448869_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66242_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66242_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400011424.mm.1 | NM_025440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011424.mm.1 | NM_025440.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011424.mm.1 | TC1400001535.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400011423.mm.1 | NM_025440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011423.mm.1 | NM_025440.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011423.mm.1 | TC1400001535.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400011425.mm.1 | NM_025440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011425.mm.1 | NM_025440.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011425.mm.1 | TC1400001535.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400011426.mm.1 | NM_025440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011426.mm.1 | NM_025440.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400011426.mm.1 | TC1400001535.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448869_PM_a_at | - | - |
Mouse Expression Set 430 | 1448869_a_at | - | - |
Murine Genome U74v2 Set | 94866_PM_at | - | - |
Murine Genome U74v2 Set | 94866_at | - | - |
Mouse Genome 430A 2.0 Array | 1448869_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1448869_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448869_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448869_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 897920 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2510390 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2510390 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002510390 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619671 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2619671 | - | - |
mouse-6 v1.1 (Illumina) | scl066242.2_14-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2510390 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066242.2_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510390 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2619671 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619671 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066242.2_14-S | - | - |
