Assay Details
| Gene Symbol: Dimt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025447.4 | NP_079723.1 | 11 - 12 | 971 | 143 | |
| GenBank mRNA | AK007759.1 | - | 11 - 12 | 923 | 143 | |
| AK011544.1 | - | 11 - 12 | 928 | 143 | ||
| AK015145.1 | - | 11 - 12 | 920 | 143 | ||
| AK078186.1 | - | 9 - 10 | 1765 | 143 | ||
| AK078401.1 | - | 9 - 10 | 1787 | 143 | ||
| AK146188.1 | - | 11 - 12 | 942 | 143 | ||
| AK161333.1 | - | 11 - 12 | 923 | 143 | ||
| BC019799.1 | - | 11 - 12 | 944 | 5053500 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
rRNA modification
rRNA processing
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
RNA binding
methyltransferase activity
rRNA methyltransferase activity
transferase activity
poly(A) RNA binding
18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase activity
rRNA modification
rRNA processing
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
RNA binding
methyltransferase activity
rRNA methyltransferase activity
transferase activity
poly(A) RNA binding
18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453461 | AF102540.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453461 | BC119308.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453461 | BC119334.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453461 | NM_146964.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117535 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118537 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163490 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338611 | AK146188.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338611 | AK161333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338611 | AK015145.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338611 | AK078186.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338611 | NM_025447.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09530883 | - | - |
| Whole Mouse Genome G4122A | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | BC019799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK078401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK146188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK161333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537381 | AK015145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338611 | AK146188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338611 | AK161333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338611 | AK015145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338611 | AK078186.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338611 | NM_025447.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100588 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135876 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09530883 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10823813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10823808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32903777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32903864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32903863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32903801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32903812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10407046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407047 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426291_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426290_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407042 | - | - |
| Mouse Genome 430 2.0 Array | 1426290_at | - | - |
| Mouse Genome 430 2.0 Array | 1426290_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426291_at | - | - |
| Mouse Genome 430 2.0 Array | 1426291_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66254_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66254_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300007554.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007554.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007554.mm.1 | TC1300001190.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300007555.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007555.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007555.mm.1 | TC1300001190.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300007556.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007556.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007556.mm.1 | TC1300001190.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300007553.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007553.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007553.mm.1 | TC1300001190.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300007552.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007552.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007552.mm.1 | TC1300001190.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300007557.mm.1 | NM_025447.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007557.mm.1 | NM_025447.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007557.mm.1 | TC1300001190.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426290_at | - | - |
| Mouse Expression Set 430 | 1426291_PM_at | - | - |
| Mouse Expression Set 430 | 1426291_at | - | - |
| Mouse Expression Set 430 | 1426290_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426290_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426290_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426291_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426291_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426290_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426291_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426291_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426290_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 435967 | - | - |
| ABI Mouse Genome Survey Microarray | 475408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6420215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6420215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734408 | - | - |
| mouse-6 v1.1 (Illumina) | scl066254.12_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066254.12_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066254.12_0-S | - | - |

Dimt1
Genomic Map