Assay Details
| Gene Symbol: Uqcrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025641.3 | NP_079917.1 | 3 - 4 | 342 | 144 | |
| GenBank mRNA | AK003266.1 | - | 3 - 4 | 307 | 144 | |
| AK003539.1 | - | 3 - 4 | 285 | 144 | ||
| AK011734.1 | - | 704 | 144 | |||
| AK012552.1 | - | 3 - 4 | 320 | 144 | ||
| AK018712.1 | - | 3 - 4 | 320 | 144 | ||
| AK054005.1 | - | 3 - 4 | 308 | 144 | ||
| AK088185.1 | - | 3 - 4 | 307 | 144 | ||
| BC011388.1 | - | 3 - 4 | 286 | 3597862 | 144 | |
| BY702808.1 | - | 3 - 4 | 315 | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23227557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23227556 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052470 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965427 | AK018712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965427 | AK054005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965427 | AK088185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965427 | BC011388.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965427 | BY702808.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139507 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108333 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108227 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052470 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965427 | AK018712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965427 | AK054005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965427 | AK088185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965427 | BC011388.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965427 | BY702808.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431852 | AK018712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431852 | AK019085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431852 | AK028137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431852 | AK054005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431852 | AK088185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P541875 | AK018712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P541875 | AK054005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P541875 | AK088185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P541875 | BY138998.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P541875 | BY702808.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23227531 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108333 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108227 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05367906 | - | - |
| Whole Mouse Genome G4122A | A_51_P431852 | AK018712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431852 | AK019085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431852 | AK028137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431852 | AK054005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431852 | AK088185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541875 | AK018712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541875 | AK054005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541875 | AK088185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541875 | BY138998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541875 | BY702808.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108227 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03678976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03678968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03678980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03678977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03678974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23227556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23227557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03678976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03988076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03294811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03993109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23227549 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432557 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452133_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453229_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432556 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515253 | - | - |
| Mouse Genome 430 2.0 Array | 1453229_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452133_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453229_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452133_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66576_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66576_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400026529.mm.1 | TC0400003330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026529.mm.1 | NM_025641.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026529.mm.1 | NM_025641.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026535.mm.1 | TC0400003330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026535.mm.1 | NM_025641.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026535.mm.1 | NM_025641.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026538.mm.1 | TC0400003330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026538.mm.1 | NM_025641.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026538.mm.1 | NM_025641.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026530.mm.1 | TC0400003330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026530.mm.1 | NM_025641.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026530.mm.1 | NM_025641.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026528.mm.1 | TC0400003330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026528.mm.1 | NM_025641.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026528.mm.1 | NM_025641.3 | Refseq |
| Mouse Expression Set 430 | 1452133_at | - | - |
| Mouse Expression Set 430 | 1452133_PM_at | - | - |
| Murine Genome U74v2 Set | 99115_PM_at | - | - |
| Murine Genome U74v2 Set | 99115_at | - | - |
| Murine Genome U74v2 Set | 169360_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169360_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452133_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452133_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452133_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452133_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453229_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453229_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 585648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4210463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2512849 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2512849 | - | - |
| mouse-6 v1.1 (Illumina) | scl066576.4_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066576.4_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210463 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2512849 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2512849 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066576.4_1-S | - | - |

Uqcrh
Genomic Map