Assay Details
| Gene Symbol: Fundc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026126.4 | NP_080402.3 | 4 - 5 | 409 | 81 | |
| GenBank mRNA | AK013460.1 | - | 4 - 5 | 387 | 81 | |
| AK014705.1 | - | 4 - 5 | 386 | 81 | ||
| AK031214.1 | - | 4 - 5 | 386 | 81 | ||
| AY234858.1 | - | 4 - 5 | 375 | 81 | ||
| BC042794.1 | - | 3 - 4 | 366 | 81 | ||
| BC049566.1 | - | 4 - 5 | 384 | 6742356 | 81 | |
| BC061464.1 | - | 4 - 5 | 379 | 6819539 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133928 | AK031214.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133928 | BC042794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133928 | BC049566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133928 | BC061464.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133928 | NM_026126.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122909 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111574 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133928 | AK031214.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133928 | BC042794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133928 | BC049566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133928 | BC061464.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133928 | NM_026126.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32471438 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111574 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09333418 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03699154 | - | - |
| Whole Mouse Genome G4122A | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168338 | AK031214.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168338 | NM_026126.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133928 | AK031214.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133928 | BC042794.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133928 | BC049566.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133928 | BC061464.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133928 | NM_026126.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32471402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15225020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14776091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14693978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32471441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600507 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428316_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460434_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428317_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600504 | - | - |
| Mouse Genome 430 2.0 Array | 1460434_at | - | - |
| Mouse Genome 430 2.0 Array | 1428316_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428316_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1460434_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67391_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67391_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00005125.mm.1 | NM_026126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005125.mm.1 | NM_026126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005125.mm.1 | TC0X00000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005128.mm.1 | NM_026126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005128.mm.1 | NM_026126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005128.mm.1 | TC0X00000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005127.mm.1 | NM_026126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005127.mm.1 | NM_026126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005127.mm.1 | TC0X00000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005123.mm.1 | NM_026126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005123.mm.1 | NM_026126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005123.mm.1 | TC0X00000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005129.mm.1 | NM_026126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005129.mm.1 | NM_026126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005129.mm.1 | TC0X00000741.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460434_at | - | - |
| Mouse Expression Set 430 | 1428317_at | - | - |
| Mouse Expression Set 430 | 1428316_a_at | - | - |
| Mouse Expression Set 430 | 1428316_PM_a_at | - | - |
| Mouse Expression Set 430 | 1460434_PM_at | - | - |
| Mouse Expression Set 430 | 1428317_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460434_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460434_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428316_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428316_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428317_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428317_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460434_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460434_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428316_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428316_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 730822 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603439 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237411 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603439 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237411 | - | - |
| mouse-6 v1.1 (Illumina) | scl067391.5_27-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067391.2_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770041 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067391.5_27-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067391.2_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550133 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715629 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2603439 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2715629 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2603439 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067391.2_52-S | - | - |

Fundc2
Genomic Map